SY3H3D6810DB Synthonix Corporation

N-((4-Methoxybenzyl)oxy)-2-nitrobenzenesulfonamide

  • MDL number No MDL found
  • Cas Number : 1384122-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57638.html

Safety Information

MSDS https://synthonix.com/57638.html
Availability details for
Quantity
  • Availability
  • Add to Cart