SY3H3D6810DC Synthonix Corporation

Methyl 2-(2-hydroxy-4,5-dimethoxybenzamido)thiazole-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 877997-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57639.html

Safety Information

MSDS https://synthonix.com/57639.html
Availability details for
Quantity
  • Availability
  • Add to Cart