SY3H3D6810DE Synthonix Corporation

N-(4-Methoxybenzyl)aniline

  • MDL number No MDL found
  • Cas Number : 3526-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57669.html

Safety Information

MSDS https://synthonix.com/57669.html
Availability details for
Quantity
  • Availability
  • Add to Cart