SY3H3D6810E0 Synthonix Corporation

5-(4-Methoxy-3-methylphenyl)-3-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 2109802-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57681.html

Safety Information

MSDS https://synthonix.com/57681.html
Availability details for
Quantity
  • Availability
  • Add to Cart