SY3H3D6810E1 Synthonix Corporation

2-(2-(2-Methyl-1,3-dioxolan-2-yl)ethyl)isoindoline-1,3-dione

  • Cas Number : 84764-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57684.html

Safety Information

MSDS https://synthonix.com/57684.html
Availability details for
Quantity
  • Availability
  • Add to Cart