SY3H3D6810E6 Synthonix Corporation

Methyl 1-benzyl-1,2,3,6-tetrahydropyridine-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 80845-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57735.html

Safety Information

MSDS https://synthonix.com/57735.html
Availability details for
Quantity
  • Availability
  • Add to Cart