SY3H3D6810E9 Synthonix Corporation

3-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazol-2(3H)-one

  • MDL number No MDL found
  • Cas Number : 1016641-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57795.html

Safety Information

MSDS https://synthonix.com/57795.html
Availability details for
Quantity
  • Availability
  • Add to Cart