SY3H3D6810EA Synthonix Corporation

Methyl 2-(4-bromophenyl)-2-((tert-butoxycarbonyl)amino)acetate

  • MDL number No MDL found
  • Cas Number : 709665-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/57796.html

Safety Information

MSDS https://synthonix.com/57796.html
Availability details for
Quantity
  • Availability
  • Add to Cart