SY3H3D6810EB Synthonix Corporation

4-(4-Methoxy-2,3,6-trimethylphenyl)but-3-en-2-one

  • MDL number No MDL found
  • Cas Number : 54757-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57807.html

Safety Information

MSDS https://synthonix.com/57807.html
Availability details for
Quantity
  • Availability
  • Add to Cart