SY3H3D6810EC Synthonix Corporation

Methyl 2-cyclopentyl-2-hydroxy-2-phenylacetate

  • MDL number No MDL found
  • Cas Number : 19833-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57813.html

Safety Information

MSDS https://synthonix.com/57813.html
Availability details for
Quantity
  • Availability
  • Add to Cart