SY3H3D6810ED Synthonix Corporation

2-(3-Methoxy-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number No MDL found
  • Cas Number : 479411-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57815.html

Safety Information

MSDS https://synthonix.com/57815.html
Availability details for
Quantity
  • Availability
  • Add to Cart