SY3H3D6810EE Synthonix Corporation

Methyl 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number No MDL found
  • Cas Number : 334018-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/57818.html

Safety Information

MSDS https://synthonix.com/57818.html
Availability details for
Quantity
  • Availability
  • Add to Cart