SY3H3D6810F0 Synthonix Corporation

Methyl 4-amino-1-(4-chlorobenzyl)piperidine-4-carboxylate dihydrochloride

  • MDL number No MDL found
  • Cas Number : 1210494-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57827.html

Safety Information

MSDS https://synthonix.com/57827.html
Availability details for
Quantity
  • Availability
  • Add to Cart