SY3H3D6810F3 Synthonix Corporation

(6bR,10aS)-3-Methyl-2,3,6b,7,8,9,10,10a-octahydro-1H-pyrido[3',4':4,5]pyrrolo[1,2,3-de]quinoxaline

  • MDL number No MDL found
  • Cas Number : 313368-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57833.html

Safety Information

MSDS https://synthonix.com/57833.html
Availability details for
Quantity
  • Availability
  • Add to Cart