SY3H3D6810F9 Synthonix Corporation

2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

  • MDL number No MDL found
  • Cas Number : 1431542-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57874.html

Safety Information

MSDS https://synthonix.com/57874.html
Availability details for
Quantity
  • Availability
  • Add to Cart