SY3H3D6810FB Synthonix Corporation

Methyl 3-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number No MDL found
  • Cas Number : 850689-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/57902.html

Safety Information

MSDS https://synthonix.com/57902.html
Availability details for
Quantity
  • Availability
  • Add to Cart