SY3H3D6810FD Synthonix Corporation

Methyl 3-(3-(tert-butyl)-4-hydroxyphenyl)propanoate

  • Cas Number : 36837-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57910.html

Safety Information

MSDS https://synthonix.com/57910.html
Availability details for
Quantity
  • Availability
  • Add to Cart