SY3H3D6810FF Synthonix Corporation

Methyl N-Boc-4-piperidinepropionate

  • MDL number No MDL found
  • Cas Number : 162504-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57988.html

Safety Information

MSDS https://synthonix.com/57988.html
Availability details for
Quantity
  • Availability
  • Add to Cart