SY3H3D681100 Synthonix Corporation

(4-(4-Methylpiperazin-1-yl)phenyl)methanamine 2,2,2-trifluoroacetate

  • MDL number MFCD28144965
  • Cas Number : 1632286-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58071.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58071.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart