SY3H3D681101 Synthonix Corporation

(R)-Methyl 2-(pyrrolidin-3-yloxy)acetate 4-methylbenzenesulfonate

  • Cas Number : 1965314-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58073.html

Safety Information

MSDS https://synthonix.com/58073.html
Availability details for
Quantity
  • Availability
  • Add to Cart