SY3H3D681103 Synthonix Corporation

Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58087.html

Safety Information

MSDS https://synthonix.com/58087.html
Availability details for
Quantity
  • Availability
  • Add to Cart