SY3H3D68110A Synthonix Corporation

5-(6-Methoxynaphthalen-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1257877-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/58153.html

Safety Information

MSDS https://synthonix.com/58153.html
Availability details for
Quantity
  • Availability
  • Add to Cart