SY3H3D68110C Synthonix Corporation

(E)-Methyl 2-(2-((6-chloropyrimidin-4-yl)oxy)phenyl)-3-methoxyacrylate

  • MDL number No MDL found
  • Cas Number : 131860-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58177.html

Safety Information

MSDS https://synthonix.com/58177.html
Availability details for
Quantity
  • Availability
  • Add to Cart