SY3H3D68110E Synthonix Corporation

2-((4-Methoxyphenyl)ethynyl)aniline

  • Cas Number : 157869-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58186.html

Safety Information

MSDS https://synthonix.com/58186.html
Availability details for
Quantity
  • Availability
  • Add to Cart