SY3H3D68110F Synthonix Corporation

5-Methyl-5H-dibenzo[b,d]azepin-6(7H)-one

  • MDL number No MDL found
  • Cas Number : 209984-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58188.html

Safety Information

MSDS https://synthonix.com/58188.html
Availability details for
Quantity
  • Availability
  • Add to Cart