SY3H3D681110 Synthonix Corporation

Methyl 4-(phenylcarbamoyl)benzoate

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58199.html

Safety Information

MSDS https://synthonix.com/58199.html
Availability details for
Quantity
  • Availability
  • Add to Cart