SY3H3D681111 Synthonix Corporation

6-Methoxy-2-(p-tolyl)benzo[d]thiazole

  • MDL number MFCD17211091
  • Cas Number : 101078-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Off-white or gray to light yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58209.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58209.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart