SY3H3D681113 Synthonix Corporation

2'-Methoxy-2-phenylacetophenone

  • MDL number No MDL found
  • Cas Number : 33470-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58230.html

Safety Information

MSDS https://synthonix.com/58230.html
Availability details for
Quantity
  • Availability
  • Add to Cart