SY3H3D681115 Synthonix Corporation

Methyl 4'-(hydroxymethyl)-[1,1'-biphenyl]-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 393522-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58242.html

Safety Information

MSDS https://synthonix.com/58242.html
Availability details for
Quantity
  • Availability
  • Add to Cart