SY3H3D68111A Synthonix Corporation

4-(4'-Methyl-[2,2'-bipyridin]-4-yl)butanoic acid

  • MDL number No MDL found
  • Cas Number : 114527-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58270.html

Safety Information

MSDS https://synthonix.com/58270.html
Availability details for
Quantity
  • Availability
  • Add to Cart