SY3H3D681122 Synthonix Corporation

Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzoate

  • Cas Number : 1150271-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58331.html

Safety Information

MSDS https://synthonix.com/58331.html
Availability details for
Quantity
  • Availability
  • Add to Cart