SY3H3D68112B Synthonix Corporation

Methyl 2-(4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate

  • MDL number MFCD12026092
  • Cas Number : 944317-66-2
  • Molecular Weight : 294.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/58385.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/58385.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart