SY3H3D68112F Synthonix Corporation

Methyl 2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number No MDL found
  • Cas Number : 478375-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58424.html

Safety Information

MSDS https://synthonix.com/58424.html
Availability details for
Quantity
  • Availability
  • Add to Cart