SY3H3D681133 Synthonix Corporation

3-Methoxy-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-amine

  • MDL number No MDL found
  • Cas Number : 457065-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58455.html

Safety Information

MSDS https://synthonix.com/58455.html
Availability details for
Quantity
  • Availability
  • Add to Cart