SY3H3D681134 Synthonix Corporation

2-Methylpropan-2-amine (S)-3,3-dimethyl-2-((((1R,2R)-2-(pent-4-yn-1-yl)cyclopropoxy)carbonyl)amino)butanoate

  • MDL number No MDL found
  • Cas Number : 1425038-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58460.html

Safety Information

MSDS https://synthonix.com/58460.html
Availability details for
Quantity
  • Availability
  • Add to Cart