SY3H3D681136 Synthonix Corporation

4-Methoxy-3-(3-(4-methylpiperazin-1-yl)propoxy)aniline

  • MDL number No MDL found
  • Cas Number : 846023-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58486.html

Safety Information

MSDS https://synthonix.com/58486.html
Availability details for
Quantity
  • Availability
  • Add to Cart