SY3H3D681139 Synthonix Corporation

2-((4-Methoxyphenyl)ethynyl)benzaldehyde

  • Cas Number : 176910-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58593.html

Safety Information

MSDS https://synthonix.com/58593.html
Availability details for
Quantity
  • Availability
  • Add to Cart