SY3H3D68113A Synthonix Corporation

Methyl 7-(5-fluoro-2-methoxyphenyl)-2-(methylsulfonyl)thieno[3,2-d]pyrimidine-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 1462950-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58599.html

Safety Information

MSDS https://synthonix.com/58599.html
Availability details for
Quantity
  • Availability
  • Add to Cart