SY3H3D68113B Synthonix Corporation

5-Methyl-3,4-diphenylisoxazole

  • MDL number No MDL found
  • Cas Number : 37928-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58603.html

Safety Information

MSDS https://synthonix.com/58603.html
Availability details for
Quantity
  • Availability
  • Add to Cart