SY3H3D68113F Synthonix Corporation

Methyl 4-(2-chloro-4-hydroxystyryl)benzoate

  • Cas Number : 1268246-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58616.html

Safety Information

MSDS https://synthonix.com/58616.html
Availability details for
Quantity
  • Availability
  • Add to Cart