SY3H3D681140 Synthonix Corporation

2-Methyl-4-phenyl-1H-indene

  • MDL number No MDL found
  • Cas Number : 159531-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58619.html

Safety Information

MSDS https://synthonix.com/58619.html
Availability details for
Quantity
  • Availability
  • Add to Cart