SY3H3D681146 Synthonix Corporation

N-(3-Methoxyphenyl)cinnamamide

  • Cas Number : 127033-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58654.html

Safety Information

MSDS https://synthonix.com/58654.html
Availability details for
Quantity
  • Availability
  • Add to Cart