SY3H3D681148 Synthonix Corporation

2-Methyl-4-(2-methylbenzamido)benzoic acid

  • MDL number No MDL found
  • Cas Number : 317374-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58659.html

Safety Information

MSDS https://synthonix.com/58659.html
Availability details for
Quantity
  • Availability
  • Add to Cart