SY3H3D68114A Synthonix Corporation

5-Methoxy-4-((4-methoxybenzyl)oxy)-2-nitrobenzoic acid

  • MDL number No MDL found
  • Cas Number : 1643979-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58667.html

Safety Information

MSDS https://synthonix.com/58667.html
Availability details for
Quantity
  • Availability
  • Add to Cart