SY3H3D68114D Synthonix Corporation

Methyl 5-chloro-1H-indole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 172595-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53759.html

Safety Information

MSDS https://synthonix.com/53759.html
Availability details for
Quantity
  • Availability
  • Add to Cart