SY3H3D681151 Synthonix Corporation

Methyl 2-amino-4-(benzyloxy)-5-methoxybenzoate

  • MDL number No MDL found
  • Cas Number : 61032-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58697.html

Safety Information

MSDS https://synthonix.com/58697.html
Availability details for
Quantity
  • Availability
  • Add to Cart