SY3H3D681153 Synthonix Corporation

2-Methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine

  • MDL number No MDL found
  • Cas Number : 1026-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58702.html

Safety Information

MSDS https://synthonix.com/58702.html
Availability details for
Quantity
  • Availability
  • Add to Cart