SY3H3D681158 Synthonix Corporation

Methyl N-Cbz-4-piperidineacetate

  • Cas Number : 170737-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58757.html

Safety Information

MSDS https://synthonix.com/58757.html
Availability details for
Quantity
  • Availability
  • Add to Cart