SY3H3D681159 Synthonix Corporation

Methyl 2-(4-bromobenzyl)-3-((tert-butoxycarbonyl)amino)propanoate

  • Cas Number : 886366-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58787.html

Safety Information

MSDS https://synthonix.com/58787.html
Availability details for
Quantity
  • Availability
  • Add to Cart