SY3H3D68115C Synthonix Corporation

Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoate

  • Cas Number : 890839-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58819.html

Safety Information

MSDS https://synthonix.com/58819.html
Availability details for
Quantity
  • Availability
  • Add to Cart